Cortisone 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 17alpha,21-Dihydroxypregn-4-ene-3,11,20-trione

460520010 460522500
ACRO460520010EA 34 GBP
ACRO460520010 ACRO460522500
Cortisone 98%
Cortisone
Formula: C₂₁H₂₈O₅
MW: 360.44 g/mol
Melting Pt: 223…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00003610
CAS Number: 53-06-5

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 223 to 228 °C
Loss on drying ≤2 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR