Self Seal Sterilisation Pouches
Supplier: DISTRIBUTOR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WPSS090200 WPSS190330 WPSS305381 WPSS135255 WPSS090230
DIUKWPSS090200EA 389 GBP
DIUKWPSS090200 DIUKWPSS190330 DIUKWPSS305381 DIUKWPSS135255 DIUKWPSS090230
Self Seal Sterilisation Pouches
Bags Sterilisation Pouches
These pouches are manufactured from tinted blue, multi-layer PET/PP film. It features a self-adhesive strip which by removing the backing strip and folding over it's possible to create and integral seal that would withstand sterilisation.
  • Single use
  • Chevron shaped end with easy opening
  • High strength and good barrier properties with a soft texture
  • Suitable for ETO sterilisation and formaldehyde
  • Printed with Class 1 process indicators compliant with ISO.11140-1
  • Manufactured under BS EN ISO.9001.2008 and BS EN ISO 13485:2003
Certifications: Conforms to BS EN ISO11607-1:2006 part 1 and BS EN 868-3 and 5:2009. Sterilisation indicators comply with BS EN ISO 11140-1:2005.