4-Nitrophenylphosphorylcholine ≥97%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J66465.MC
J66465.ME
J66465.03
J66465.MCEA
44.9
GBP
J66465.MC
J66465.ME
J66465.03
4-Nitrophenylphosphorylcholine ≥97%
4-Nitrophenylphosphorylcholine
O-(4-Nitrophenylphosphoryl)choline is a useful probe in the assay of phospholipase C.
- Soluble in water
Caution: Moisture and light sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Formula:
C₁₁H₁₇N₂O₆P MW: 304.239 g/mol Melting Pt: 233…235 °C Storage Temperature: Freezer |
MDL Number:
MFCD00077834 CAS Number: 21064-69-7 |
Specification Test Results
Thin layer chromatography | 98.5% min |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...