Carbetapentane citrate, white ≥95% (by HPLC), powder
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
J64446.MF
J64446.03
J64446.MFEA
29.3
GBP
J64446.MF
J64446.03
Carbetapentane citrate, white ≥95% (by HPLC), powder
Carbetapentane citrate
Sigma receptor agonist.
Formula:
C₂₆H₃₉NO₁₀ Storage Temperature: Ambient |
MDL Number:
MFCD00055697 CAS Number: 23142-01-0 EINECS: 245-449-5 Merck Index: 14,1794 |
Specification Test Results
Colour | White |
Form | Powder |
Assay (HPLC) | 95,0% min |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...