N-Acetyl-Asp-Glu
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A5930-100MG
A5930-25MG
SIALA5930-100MGEA
455
GBP
SIALA5930-100MG
SIALA5930-25MG
N-Acetyl-Asp-Glu
Proteins and Peptides
Endogenous neurotransmitter localised to neurons with high affinity for metabotropic glutamate receptors, mGluR₃. It is an antagonist at NMDA receptors. Catabolised by carboxypeptidase II, which is expressed on astrocyte membranes, to N-acetylaspartate and glutamate.
- Optical activity: [α]22/D −34,5°, c = 1,1 in H₂O (lit.)
- Solubility: Soluble 50 mg/ml (H₂O)
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...