Glove boxes, nitrogen dry chambers 850 series
Supplier: PLASLABS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
850-NB/EXP 850-NBB/EXP
135-0667EA 0 GBP
135-0667 135-0668
Glove boxes, nitrogen dry chambers 850 series
Glove Boxes and Glove Bags
The nitrogen dry boxes are completely enclosed chambers which are ideal for working in a preferred atmosphere. Inert gases like argon, nitrogen, and plasma are easily maintained. They are compact, portable, and lightweight. The design makes them easily moveable from lab to lab. Everything that is required is included with the units. All you need is the gas of choice and research or production materials.
  • Bright fluorescent light system
  • Front sloped viewing panel minimises glare
  • Adjustable vacuum gauge on transfer chamber
  • Four ground key cock valves for
More Product Information