Glove boxes, Basic 818 series chamber
Supplier: PLASLABS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
818-GB/EXP 818-GBB/EXP
135-0662EA 0 GBP
135-0662 135-0663
Glove boxes, Basic 818 series chamber
Glove Boxes and Glove Bags
These basic glove boxes are engineered to fit general laboratory isolation applications. They can be easily modified for specific uses. Units are very handy for isolating sensitive research studies from a hostile exterior environment.
  • Coved, easy to clean corners with no hidden dark spots
  • Double layered, closed cellular neoprene gasketing system
  • Stainless steel 'O' rings for gloves and ambidextrous, white Hypalon gloves for superior chemical and UV resistance
  • Adjustable vacuum gauge on transparent transfer chamber, 30 long×28 cm diameter (Int.Ø)
  • Four ground key-cock valves for purging and a levelling tray for transferring liquids through the transfer chamber
  • All clamps fully adjustable to compensate for wear
These economical units are compact, portable, lightweight and self-contained. They consist of an optically clear, one piece acrylic top and a die moulded thermoset, bright white bottom (819-GB only).
Hospital grade multiple electrical outlet strip.