Compact glove boxes, 830 Series (powder weighing)
Supplier: PLASLABS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
830-ABB/EXP 830-ABC/EXP 830-ABD/EXP
135-0664EA 0 GBP
135-0664 135-0665 135-0666
Compact glove boxes, 830 Series (powder weighing)
Glove Boxes and Glove Bags
The 830 minimises drafts and maximises effectiveness of your balance. This unit features cleanliness, safety, complete containment, portability, and a draft-free atmosphere. The compact glove box is designed specifically for use in hazardous situations where materials to be weighed must be contained.
  • Top section is removable for easy installation of equipment
  • Four purging gas valves (two on the transfer chamber and two on the main chamber)
  • HEPA and organic vapour canister filters (replaceable)
  • Hospital grade multiple electrical outlet strip
  • White levelling tray for easy transferring of liquids
  • All clamps are stainless steel and completely adjustable
  • White ambidextrous Hypalon gloves for superior chemical UV resistance
  • Pressure relief valve minimises glove 'fight back'
The chamber is ideal for determining the dry weight or moisture content of aqueous solutions, adhesives, cereals, toxic chemicals, pharmaceuticals, paper, plastics, and light radioactive materials. It minimises product degradation caused by moisture absorption. The 830 is beneficial when working with toxic substances, asbestos fibres, sewage residue, and harmful liquid vapours. For use with analytical balances, top pan balances and/or moisture analysers.
Transfer chamber: 30 long×28 cm diameter (Int.Ø)
Includes three polycarbonate canisters filled with DesiSphere™ medium, hose connections, and diaphragm pump mounted on 0,375.
Accessories information: Workstation Ioniser: Effectively eliminates static charges which exist within the chamber. Pulse rate (adjustable from 1 to 5 pulses/ sec.). Ion balance controls allow for calibration and adjustment of the ioniser to give optimum ion balance and output. Compact design utilises minimum amount of valuable work space. Effective ion coverage range up to 36.
800-DT - Drying Train Package with Pump: Includes three polycarbonate canisters filled with DesiSphere™ medium, hose connections, and diaphragm pump mounted on 0,375.