Water, distilled

Supplier: DISTRIBUTOR
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Hydrogen oxide

WA11600-275 WA116600-355
DIUKWA11600-275EA 25.6 GBP
DIUKWA11600-275 DIUKWA116600-355
Water, distilled
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR