Dialysis tubing
Supplier: BIODESIGN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D100 D102 D104 D106 D108
511-0722EA 417 GBP
511-0722 511-0723 511-0724 511-0725 511-0726
Dialysis tubing
Tubing Dialysis Tubing
Dialysis tubing are seamless, thin, transparent and made of cellulose.
Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies. Supplied in an airtight, moisture-proof container for maximum shelf life.