Human recombinant RANTES (from E. coli)

Supplier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ORB49268-1 ORB49268-10 ORB49268-50
BIRBORB49268-1EA 0 GBP
BIRBORB49268-1 BIRBORB49268-10 BIRBORB49268-50
Human recombinant RANTES (from E. coli)
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name RANTES
Environmentally Preferable
Protein synonyms Beta chemokine RANTES
Protein/peptide type Recombinant
Species Human
Source E. coli
Conjugation Unconjugated
Sequence SPYSSDTTPCCFAYIARPLPRAHIKEYFYTSGKCSNPAVVFVTRKNRQVCANPEKKWVREYINSL EMS
UniProtKB P13501 (Human)
Purity >95% as determined by reducing SDS-PAGE.
Formulation Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4.
Storage conditions Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
Tested applications SDS-PAGE

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR