Autosampler vial convenience kits
Supplier: Restek
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21192 21193 21194 21195 24638 24639 24640 24641
REST21192EA 105 GBP
REST21192 REST21193 REST21194 REST21195 REST24638 REST24639 REST24640 REST24641
Autosampler vial convenience kits
Vials
Convenience kits with black PP open-hole caps and 8 mm red PTFE/silicone septa.
Packaging: Vials are packaged in a clear-lid tray. Preassembled caps with septa are packaged separately in a plastic bag.