Anti-TUB Rat Monoclonal Antibody [clone: YL1/2]

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NB600-506 NB600-506SS
NOVUNB600-506EA 618 GBP
NOVUNB600-506 NOVUNB600-506SS
Anti-TUB Rat Monoclonal Antibody [clone: YL1/2]
Antibodies
Rat Monoclonal Tubulin Antibody (YL1/2). Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin, Immunoprecipitation, Radioimmunoassay. Tested Reac

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: Tubulin
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol TUB
Environmentally Preferable
Antigen name Tubulin
Antigen synonyms B-ALPHA-1|TUBA3|LIS3
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone YL1/2
Reactivity Human
Host Rat
Gene ID 7846
Isotype IgG2a
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR