Anti-RBFOX3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

NBP1-77686 NBP1-77686SS
NOVUNBP1-77686EA 677 GBP
NOVUNBP1-77686 NOVUNBP1-77686SS
Anti-RBFOX3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal RBFOX3/NeuN Antibody. Tested Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: RBFOX3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol RBFOX3
Antigen name RNA Binding Protein, Fox-1 Homolog 3
Environmentally Preferable
Antigen synonyms FOX-3|FOX3|NEUN|HRNBP3
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 146713
Isotype
ImmunoChemistry Yes
ImmunoFluorescence Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR