Anti-DPP4 Mouse Monoclonal Antibody (CF594) [clone: DPP4/910]

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNC940910-500 BNC940910-100
BNC940910-500EA 533 GBP
BNC940910-500 BNC940910-100
Anti-DPP4 Mouse Monoclonal Antibody (CF594) [clone: DPP4/910]
Antibodies
Recognizes a glycoprotein of 110 kDa, identified as CD26. It is an atypical serine protease belonging to the prolyl oligopeptidase family. It is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection.

Type: Primary
Antigen: DPP4
Clonality: Monoclonal
Clone: DPP4/910
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR