Anti-ECM1 Mouse Monoclonal Antibody (CF640R) [clone: ECM1/792]

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNC400792-500 BNC400792-100
BNC400792-500EA 533 GBP
BNC400792-500 BNC400792-100
Anti-ECM1 Mouse Monoclonal Antibody (CF640R) [clone: ECM1/792]
Antibodies
This MAb reacts with a reduction-resistant epitope present in both free and SIgA bound Secretory Component. It does not react with the cell lines lacking secretory component. The antibody is useful for studying the distribution and level of both free and bound secretory component. Secretory component is differentially expressed in epithelium, and the antibody is a popular marker for identifying subpopulations of epithelial cells and epithelial differentiation. The Secretory component antibody is a useful research tool for studying mucosal immunity, inflammation, remodeling, differentiation and tumorigenesis, all processes associated with differential secretory component expression.

Type: Primary
Antigen: ECM1
Clonality: Monoclonal
Clone: ECM1/792
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR