Anti-Biotin Mouse Monoclonal Antibody (CF594) [clone: Hyb8]

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNC940400-500 BNC940400-100
BNC940400-500EA 533 GBP
BNC940400-500 BNC940400-100
Anti-Biotin Mouse Monoclonal Antibody (CF594) [clone: Hyb8]
Antibodies
This antibody recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. This MAb is highly specific to biotin and shows no cross-reaction with other structurally related compounds. It has a very high affinity for biotin and is excellent for use in various amplification techniques. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: Hyb8
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR