Anti-ANPEP Mouse Monoclonal Antibody (CF405S) [clone: B-F10]

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNC040343-500 BNC040343-100
BNC040343-500EA 533 GBP
BNC040343-500 BNC040343-100
Anti-ANPEP Mouse Monoclonal Antibody (CF405S) [clone: B-F10]
Antibodies
Recognizes an integral membrane glycoprotein of 150 kDa, identified as CD13 (also known as aminopeptidase-N). The CD13 antigen is present on most cells of myeloid origin including granulocytes, monocytes, mast cells, and GM-progenitor cells. It is also expressed by the majority of AML, CML in myeloid blast crisis, and in a smaller fraction of lymphoid leukemias. CD13 is absent from normal lymphocytes, platelets and erythrocytes. CD13 is also present on fibroblasts; endothelial cells, epithelial cells from renal proximal tubules and intestinal brush border, bone marrow stromal cells,More Product Information
Order Now

SPECIFICATIONS

Antigen symbol ANPEP
Environmentally Preferable
Antigen name CD13
Antigen synonyms ANPEP|PEPN|Microsomal aminopeptidase|Lap1|p150|gp150|APN|Aminopeptidase N|hAPN|Aminopeptidase M|Alanyl aminopeptidase|AP-M|Myeloid plasma membrane glycoprotein CD13
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 405S
Clone Clone B-F10
Reactivity Human
Host Mouse
Gene ID 1239
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR