Anti-MUC1 Mouse Monoclonal Antibody (CF568) [clone: GP1.4]

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNC680159-500 BNC680159-100
BNC680159-500EA 533 GBP
BNC680159-500 BNC680159-100
Anti-MUC1 Mouse Monoclonal Antibody (CF568) [clone: GP1.4]
Antibodies
In Western blotting, this antibody recognizes proteins in MW range of 265-400 kDa, identified as different glycoforms of EMA. This MAb reacts with the DTRP epitope in the tandem repeats. The α subunit has cell adhesive properties. It can act both as an adhesion and an anti-adhesion protein. EMA may provide a protective layer on epithelial cells against bacterial and enzyme attack. The β subunit contains a C-terminal domain, which is involved in cell signaling, through phosphorylations and protein-protein interactions. In immunohistochemical assays, it superbly stains routine formalin/paraffin carcinoma tissues. Antibody to EMA is useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in bone marrow or liver.

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: GP1.4
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR