Anti-FN1 Mouse Monoclonal Antibody (Purified) [clone: TV-1]

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNUB0029-500 BNUB0029-100
BNUB0029-500EA 487 GBP
BNUB0029-500 BNUB0029-100
Anti-FN1 Mouse Monoclonal Antibody (Purified) [clone: TV-1]
Antibodies
Fibronectin is a soluble dimeric glycoprotein of 440 kDa, which is present in cells, extracellular matrix, and blood. This MAb reacts with the cellular as well as plasma form of fibronectin. Reportedly, after iv administration, this MAb localizes to tumor vessels where it binds to the underlying basement. Epitope recognized by this antibody is not accessible in normal tissues to the circulating MAb indicating that it can be used to specifically target tumor vessels in vivo. TV-1 is reportedly useful for delivering vasoactive agents to tumors to induce increased vascular permeability or bloodMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol FN1
Environmentally Preferable
Antigen name Fibronectin
Antigen synonyms FN1|Ugl-Y3|FNZ|Migration stimulating factor (MSF)|GFND2|Cold insoluble globulin (CIG)|LETS|GFND|FINC
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone Clone TV-1
Reactivity Pig,Human,Rat,Mouse
Host Mouse
Gene ID 203717
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0,2 mg/ml
Storage buffer PBS/0,05% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR