Continuous monitoring of temperature (−200 to +200 °C), humidity, power and CO₂.
- 24/7 continuous monitoring
- Automatic alert: User-set parameters with alerts onsite or remotely via SMS, email or voice call
- Easy access to data via any web-enabled device
- Reporting and audit trails: Data is stored and backed-up for easy reporting and for audit purposes
- Wireless data transmission: No wiring or rebuilding required for installation
- Flexible: Supports systems of all sizes and structures. Systems can be incrementally extended and relocated
The t-Scan monitoring solution provides confidence and peace of mind through comprehensive and traceable monitoring, reporting and alerting services. Suitable for use in controlled environments such as the medical, pharmaceutical, research and food production sectors.
Where data integrity and notification services are important, t-Scan provides a compliant and market leading system. Data is transmitted over secure lines with audit trail logs for all system activity. Managed user credentials ensure multiple views and user privileges. This means that customers have flexibility in assigning the level of user functionality and monitoring points that can be viewed.
This flexible system allows monitoring of temperature, humidity, power and CO₂ as well as other metrics that can be tailored to suit the customer’s specific operating environment. Multiple user access levels control system access and alerts (email, SMS, voice, visual cues) can be set to suit each user profile.
Reporting can also be customised to suit individual/specific requirements in all standard outputs such as pdf. HTMI, CSV, .XLS formats. The easy to use software features clear, colour-coded visual displays and intuitive screens.
The support package (LOCATSCANSUPPORT) can include annual calibration and sensor checks with UKAS accredited reference probes or to UKAS 17025 standard, access to the T-Scan helpdesk and 24/7 system health checks and system notifications.
Certifications: Full IQOQ validation and CFR21 Part11 D compliance.
Ordering information: In order to ensure that your t-Scan system is configured correctly, it is essential that a full site survey is carried out. This way, our trained engineers can assess the optimal solution for your requirements. Please contact our Technical Office on 01342 337447 or techoffice.uk@Avantor.com to book an appointment.
Delivery information: A complete system will comprise: hardware for measuring and transmitting data, software drivers, access to client tscanweb site and product certifications. Commissioning, site set-up and system training are all part of installation process.
Caution: * GPRS available as an option, please enquire
** Network point required
*** Full specifications will be provided with the quotation