Cannula filter discs
Supplier: QUALITY LAB ACCESSORIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FIL010-DK FIL045-DK FIL070-DK FIL090-DK
QLAAFIL010-DKEA 601 GBP
QLAAFIL010-DK QLAAFIL045-DK QLAAFIL070-DK QLAAFIL090-DK
Cannula filter discs
Filters Filter Discs
Cannula filters for most sampling methods, specifically formulated to provide the specified micron ratings.
  • Filters are made from inert, high-density PE
  • Porous micron filters are made of ultrahigh molecular weight PE (UHMWPE)
  • All formulations have been verified by Porisometer testing