4-Nitrophenyl-β-D-xylopyranoside ≥98% (by TLC), powder, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
PNPX, 4-Nitrophenyl β-D-xylopyranoside
M-Clarity™ quality level = MQ300
N2132-1G
N2132-500MG
SIALN2132-1GEA
673
GBP
SIALN2132-1G
SIALN2132-500MG
4-Nitrophenyl-β-D-xylopyranoside ≥98% (by TLC), powder, Sigma-Aldrich®
4-Nitrophenyl-beta-D-xylopyranoside
Formula:
C₁₁H₁₃NO₇ Storage Temperature: Freezer |
MDL Number:
MFCD00047519 CAS Number: 2001-96-9 EINECS: 217-897-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...