Dimetridazole 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,2-Dimethyl-5-nitroimidazole

210330010 210331000
ACRO210330010EA 301 GBP
ACRO210330010 ACRO210331000
Dimetridazole 98%
Dimetridazole
Formula: C₅H₇N₃O₂
MW: 141.13 g/mol
Melting Pt: 139…140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00047046
CAS Number: 551-92-8

Order Now

Specification Test Results

Appearance Cream to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 137 to 142 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR