Picrotoxin (picrotoxinin + picrotin) 98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Cocculin
131210050
131210010
131210250
ACRO131210050EA
105
GBP
ACRO131210050
ACRO131210010
ACRO131210250
Picrotoxin (picrotoxinin + picrotin) 98%
Picrotoxin (picrotoxinin + picrotin)
Formula:
C₃₀H₃₄O₁₃ Melting Pt: 199…202 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00074824 CAS Number: 124-87-8 UN: 3462 ADR: 6.1,II |
Specification Test Results
Appearance | White to light yellow Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 199 to 203 °C |
HPLC | ≥97.5 % (Mixture of Picrotoxinin and Picrotin) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...