Ergosterol 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

117810050 117810250 117811000
ACRO117810050EA 42.5 GBP
ACRO117810050 ACRO117810250 ACRO117811000
Ergosterol 98%
Ergosterol
Formula: C₂₈H₄₄O
Boiling Pt: 250 °C (1.3Pa)
Melting Pt: 156…158 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003623
CAS Number: 57-87-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or needles
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation -105° to -135° (20°C, 589 nm) (c=1, CHCl3)
Nitrate (NO3) ≤5 ppm
Sulfate (SO4) ≤30 ppm
Antimony (Sb) ≤20 ppm
Arsenic (As) ≤1 ppm
Calcium (Ca) ≤20 ppm
Iron (Fe) ≤30 ppm
Magnesium (Mg) ≤20 ppm
Manganese (Mn) ≤0.5 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤250 ppm
Heavy metals (as Pb) ≤10 ppm
Volatile acids (CH3COOH) ≤0.001 %
Insoluble matter ≤0.001 %
Reducing substances Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR