VWR® Bacterial Freezing Media

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

N400-25PKEA 0 GBP
N400-25PK
VWR® Bacterial Freezing Media
Cell Freezing Media

Bacterial freezing media is a sterile, ready to use cryopreservation media for long term storage of bacterial cultures. Ideal for the preservation of transformed bacteria as well as non fastidious bacteria.


  • Each vial contains 0,5 ml LB broth (Miller formulation) with 30% glycerol
  • Vials are marked at 0,5 ml increments and provide an area for recording strain information
  • Viability of frozen cultures after thawing is 85%


Add 1 ml of saturated culture to the vial, vortex, and store at –70 °C


Delivery information: Supplied in a convenient storage box containing 25 single use, 2 ml capacity vials.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR