3,4-Diaminothiophene dihydrochloride 96%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Thiophene-3,4-diamine dihydrochloride

325890010 325890050
ACRO325890010EA 62.7 GBP
ACRO325890010 ACRO325890050
3,4-Diaminothiophene dihydrochloride 96%
3,4-Diaminothiophene dihydrochloride

Appearance: Grey to brown Crystalline powder

Formula: C₄H₆N₂S
Melting Pt: 240…245 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671511
CAS Number: 90069-81-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Grey to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥95 %
Loss on drying ≤3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR