Imject™ Bovine serum albumin (BSA), maleimide activated
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
77115
77116
PIER77115EA
611
GBP
PIER77115
PIER77116
Imject™ Bovine serum albumin (BSA), maleimide activated
Proteins and Peptides
Imject™ BSA maleimide-activated enables conjugation of sulfhydryl-containing peptide antigens to immunogenic bovine serum albumin carrier protein for immunisation and antibody production.