ɑ-Chloralose ≥90%, powder

Supplier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

0210045980 0210045910
ICNA0210045980EA 1730 GBP
ICNA0210045980 ICNA0210045910
ɑ-Chloralose ≥90%, powder
alpha-Chloralose

Soluble in ethanol (10 mg/ml - clear to slightly hazy, colourless to faintly yellow or yellow-brown).


α-Chloralose, may contain approx. 10% of β-Anomer


α-Chloralose is used as an anesthetic for laboratory animals.

Formula: C₈H₁₁Cl₃O₆
MW: 309.52 g/mol
MDL Number: MFCD00005542
CAS Number: 15879-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR