5-Iodopyridin-2-ol 97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Hydroxy-5-iodopyridine, 5-Iodopyridin-2(1H)-one
L20044.03
L20044.06
L20044.03EA
18.3
GBP
L20044.03
L20044.06
5-Iodopyridin-2-ol 97%
5-Iodopyridin-2-ol
Purity: 97%
CAS No.: 13472-79-2
Molecular Formula: C5H4INO
Formula Weight: 221.00
Melting Point: 189-191°
Beilstein Reference: 108752
MDL No.: MFCD00234058
Formula:
C₅H₄INO MW: 221 g/mol Melting Pt: 189…191 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234058 CAS Number: 13472-79-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...