trans-1-Octenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (E)-4,4,5,5-Tetramethyl-2-(oct-1-en-1-yl)-1,3,2-dioxaborolane

L19697.03 L19697.MD
L19697.03EA 48.5 GBP
L19697.03 L19697.MD
trans-1-Octenylboronic acid pinacol ester 97%
trans-1-Octenylboronic acid pinacol ester

Purity: 97% CAS No.: 83947-55-1 Molecular Formula: C14H27BO2 Formula Weight: 238.18 Refractive Index: 1.4460 Beilstein Reference: 7291389 MDL No.: MFCD03453667

Formula: C₁₄H₂₇BO₂
MW: 238.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03453667
CAS Number: 83947-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR