2-Bromomethyl-3-hydroxypyridine hydrobromide ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
L19525.03
L19525.06
L19525.03EA
104
GBP
L19525.03
L19525.06
2-Bromomethyl-3-hydroxypyridine hydrobromide ≥98%
2-Bromomethyl-3-hydroxypyridine hydrobromide
Purity: 98+%
CAS No.: 87440-88-8
Molecular Formula: C6H6BrNO.HBr
Formula Weight: 268.94
Melting Point: 191-193°
Beilstein Reference: 3699680
UN No.: UN3263
MDL No.: MFCD00466476
Formula:
C₆H₆BrNO·HBr MW: 268.94 g/mol Melting Pt: 191…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD00466476 CAS Number: 87440-88-8 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...