3-(4-Chlorophenyl)pentanedioic acid ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

L18693.14 L18693.06
L18693.14EA 0 GBP
L18693.14 L18693.06
3-(4-Chlorophenyl)pentanedioic acid ≥98%
3-(4-Chlorophenyl)pentanedioic acid

Purity: 98% CAS No.: 35271-74-0 Molecular Formula: C11H11ClO4 Formula Weight: 242.66 Melting Point: 164-168° Beilstein Reference: 1976828 MDL No.: MFCD00190249

Formula: C₁₁H₁₁ClO₄
MW: 242.6600 g/mol
Melting Pt: 164…168 °C
MDL Number: MFCD00190249
CAS Number: 35271-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR