3-Methylthiophene-2-carboxamide 99%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
L18092.06
L18092.03
L18092.06EA
106
GBP
L18092.06
L18092.03
3-Methylthiophene-2-carboxamide 99%
3-Methylthiophene-2-carboxamide
Purity: 99%
CAS No.: 76655-99-7
Molecular Formula: C6H7NOS
Formula Weight: 141.19
Melting Point: 122-125°
Beilstein Reference: 116188
MDL No.: MFCD01208055
Formula:
C₆H₇NOS MW: 141.19 g/mol Melting Pt: 122…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD01208055 CAS Number: 76655-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...