1-Bromo-3-(tert-butyldimethylsiloxy)benzene ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (3-Bromophenoxy)(tert-butyl)dimethylsilane

L17440.03 L17440.06
L17440.03EA 22.3 GBP
L17440.03 L17440.06
1-Bromo-3-(tert-butyldimethylsiloxy)benzene ≥98%
1-Bromo-3-(tert-butyldimethylsiloxy)benzene

Purity: 98+% CAS No.: 65423-56-5 Molecular Formula: C12H19BrOSi Formula Weight: 287.27 Refractive Index: 1.5090 Boiling Point: 66°/0.3mm Beilstein Reference: 2256298 MDL No.: MFCD01318106

Formula: C₁₂H₁₉BrOSi
MW: 287.27 g/mol
Boiling Pt: 66 °C (0.3 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD01318106
CAS Number: 65423-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR