4-Nitro-L-phenylalanine monohydrate 98%
Supplier: Thermo ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(S)-2-Amino-3-(4-nitrophenyl)propanoic acid monohydrate, (S)-4-Nitrophenylalanine monohydrate, H-Phe(4-NO2)-OH.H2O
Purity: 98%
CAS No.: 207591-86-4
Molecular Formula: C9H10N2O4.H2O
Formula Weight: 228.21 (210.19anhy)
Melting Point: 236-239° dec.
Beilstein Reference: 2809673
Optical Rotation: +6.5° (c=1 in 5N HCl)
MDL No.: MFCD00051221
Formula:
C₉H₁₂N₂O₅ MW: 228.20 g/mol Melting Pt: 236…239 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00051221 CAS Number: 207591-86-4 EINECS: 213-446-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...