6-Aminocoumarin hydrochloride 97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

L11168.03 L11168.06
L11168.03EA 45.1 GBP
L11168.03 L11168.06
6-Aminocoumarin hydrochloride 97%
6-Aminocoumarin hydrochloride

Purity: 97% CAS No.: 63989-79-7 Molecular Formula: C9H7NO2.HCl Formula Weight: 197.62 Melting Point: 260° dec. Beilstein Reference: 3698955 MDL No.: MFCD00082671

Formula: C₉H₇NO₂·HCl
MW: 197.62 g/mol
Melting Pt: >260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00082671
CAS Number: 63989-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR