Ampicillin trihydrate

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

J66514.14 J66514.22 J66514.36
J66514.14EA 62 GBP
J66514.14 J66514.22 J66514.36
Ampicillin trihydrate
Ampicillin trihydrate

Ampicillin is lactam antibiotic with an amino group side chain attached to the penicillin structure.


Commonly used to select for ampicillin resistance in mutated and transformed cells


White powder


Soluble in water. Practically insoluble in ethanol

Formula: C₁₆H₁₉N₃O₄S·3H₂O
Boiling Pt: 684 °C
Melting Pt: 208 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00072036
CAS Number: 7177-48-2
EINECS: 200-709-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR