Ceftazidime pentahydrate 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

J66460.03 J66460.06
J66460.03EA 104 GBP
J66460.03 J66460.06
Ceftazidime pentahydrate 95%
Ceftazidime pentahydrate

Targets primarily Gram-negative bacteria especially Pseudomonas aeruginosa unlike other third generation cephalosporins.


White to off-white powder.


Applications: Targets primarily gram negative bacteria especially Pseudomonas aeruginosa unlike other third generation cephalosporins


Sparingly soluble in aqueous solution

Formula: C₂₂H₃₂N₆O₁₂S₂
Storage Temperature: Refrigerator
MDL Number: MFCD00153936
CAS Number: 78439-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR