Cathepsin K Substrate (fluorogenic)
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J64082.MA
J64082.MB
J64082.MAEA
109
GBP
J64082.MA
J64082.MB
Cathepsin K Substrate (fluorogenic)
Cathepsin K substrate (fluorogenic)
Molecular Formula: C30H38N6O6
Formula Weight: 578.65
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White
Solubility: Soluble in DMSO
MDL No.: MFCD07780911
Applications: A sensitive fluorogenic substrate for cathepsin K
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...