Embelin
Supplier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
CAS No.: 550-24-3
Molecular Formula: C17H26O4
Formula Weight: 294.40
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Light Sensitive
Appearance: Orange
MDL No.: MFCD00016369
Applications: An inhibitor of XIAP (X-linked inhibitor-of-apoptosis protein) that contains in vivo antitumor and anti-inflammatory activity
Formula:
C₁₇H₂₆O₄ |
MDL Number:
MFCD00016369 CAS Number: 550-24-3 EINECS: 208-979-8 Merck Index: 14,3555 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...