Phosphate, 0,5M buffer solution, pH 7,2

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

J63974.AK J63974.AP
J63974.AKEA 61.6 GBP
J63974.AK J63974.AP
Phosphate, 0,5M buffer solution, pH 7,2
Buffers Biological Buffers

CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR