Carbenoxolone disodium salt ≥97%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
J63714.03
J63714.06
J63714.03EA
46
GBP
J63714.03
J63714.06
Carbenoxolone disodium salt ≥97%
Carbenoxolone disodium salt
Purity: 97+%
CAS No.: 7421-40-1
Molecular Formula: C34H48Na2O7
Formula Weight: 614.70
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White to cream
MDL No.: MFCD00079043
Applications: A triterpenoid chemopreventive agent
Formula:
C₃₄H₄₈Na₂O₇ |
MDL Number:
MFCD00079043 CAS Number: 7421-40-1 EINECS: 231-044-0 Merck Index: 14,1793 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...