Dibucaine hydrochloride

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

J62804.06 J62804.14
J62804.06EA 38.9 GBP
J62804.06 J62804.14
Dibucaine hydrochloride
Dibucaine hydrochloride

CAS No.: 61-12-1 Molecular Formula: C20H29N3O2.HCl Formula Weight: 379.92 Physical Form: Crystalline powder Appearance: White Solubility: Freely soluble in water, alcohol, acetone, or chloroform MDL No.: MFCD00012735


Applications: Potent, long-acting local anesthetic

Formula: C₂₀H₃₀ClN₃O₂
MW: 379.929 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012735
CAS Number: 61-12-1
EINECS: 200-498-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR