Evodiamine
Supplier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells.
CAS No.: 518-17-2
Molecular Formula: C19H17N3O
Formula Weight: 303.40
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: Off-white to pale yellow
MDL No.: MFCD06407824
Applications: A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells
Formula:
C₁₉H₁₇N₃O |
MDL Number:
MFCD06407824 CAS Number: 518-17-2 Merck Index: 14,3908 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...