Clomipramine hydrochloride

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

J62485.03 J62485.06
J62485.03EA 28.8 GBP
J62485.03 J62485.06
Clomipramine hydrochloride
Clomipramine hydrochloride

CAS No.: 17321-77-6 Molecular Formula: C19H23ClN2.HCl Formula Weight: 351.32 Physical Form: Powder Appearance: White to off-white Melting Point: 189-190° MDL No.: MFCD00069234


Applications: Potent, selective 5-HT uptake blocker

Formula: C₁₉H₂₄Cl₂N₂
Melting Pt: 189…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00069234
CAS Number: 17321-77-6
EINECS: 241-344-3
Merck Index: 14,2387

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR