Maprotiline hydrochloride 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

J62321.03 J62321.06
J62321.03EA 84.7 GBP
J62321.03 J62321.06
Maprotiline hydrochloride 99%
Maprotiline hydrochloride

Purity: 99% CAS No.: 10347-81-6 Molecular Formula: C20H23N.HCl Formula Weight: 313.87 Physical Form: Crystalline powder Appearance: White Melting Point: 230-232° Solubility: Soluble to 50mg/ml in water. Soluble in ethanol at 10mM Beilstein Reference: 5463242 MDL No.: MFCD00079464


Applications: Selective norepinephrine uptake inhibitor

Formula: C₂₀H₂₃N
Melting Pt: 230…232 °C
Storage Temperature: Ambient
MDL Number: MFCD00079464
CAS Number: 10347-81-6
EINECS: 233-758-8
Merck Index: 14,5748

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR