Trypsin inhibitor

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

J60982.MC J60982.03
J60982.MCEA 92.2 GBP
J60982.MC J60982.03
Trypsin inhibitor
Proteins and Peptides

CAS No.: 9035-81-8
Storage Temperature: 2 to 8 °C
Physical Form: Powder
Appearance: Off-white to pale brown
Solubility: Dissolve in 1x PBS at 1 to 10 mg/ml for stock solution
MDL No.: MFCD00082103


Applications: Inhibits the pancreatic enzyme trypsin

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR