Ifenprodil hemitartrate 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

J60932.MA J60932.MF
J60932.MAEA 80.6 GBP
J60932.MA J60932.MF
Ifenprodil hemitartrate 99%
Ifenprodil hemitartrate

Purity: 99% CAS No.: 23210-56-2 Molecular Formula: C21H27NO2.0.5C4H6O6 Formula Weight: 400.49 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD01529999


Applications: NMDA receptor antagonist, acting at the polyamine site

Formula: C₂₁H₂₇NO₂·0.5C₄H₆O₆
MDL Number: MFCD01529999
CAS Number: 23210-56-2
EINECS: 245-491-4
Merck Index: 14,4895

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR